Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Inlet liners
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
  • Liners packed with wool help to trap non-volatile material
Product Family Options
Programmed temperature split/splitless (PSS) injector liners
  • Omschrijving
    Ø uitw.
    Ø inw.
    Lengte
  • Liner/hour Glass for POC injector
    4 mm
    2,4 mm
    19,05 mm
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
    Specifications
    Cat. No.
    PERCN6101703
  • Quartz liner for splitless operation
    4 mm
    1 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
    Specifications
    Cat. No.
    PERCN6121006
  • Quartz liner for split operation
    4 mm
    2 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 41,10 €
    41,10 €
    1 * 1 ST
       41,10 €
    Specifications
    Cat. No.
    PERCN6121004
  • Siltek deactivated glass PSS liner
    4 mm
    1 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
    Specifications
    Cat. No.
    PERCN6502000
  • Siltek deactivated glass PSS liner for split operation
    4 mm
    2 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 862,00 €
    862,00 €
    1 * 1 ST
       862,00 €
    Specifications
    Cat. No.
    PERCN6502002
  • Glass injector liner
    4 mm
    2 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 809,00 €
    809,00 €
    1 * 1 ST
       809,00 €
    Specifications
    Cat. No.
    PERCN9302949
  • On-column glass liner
    4 mm
    2,4 mm
    86,2 mm
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
    Specifications
    Cat. No.
    PERCN6101539
  • Silanized Glass Wool
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
    Specifications
    Cat. No.
    PERC03300905
Capillary split/splitless injector liners
  • Omschrijving
    Ø uitw.
    Ø inw.
    Lengte
  • Clarus cup split glass liner
    6 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
    Specifications
    Cat. No.
    PERCN6502011
  • Clarus cyclosplitter glass liner
    6 mm
    4 mm
    92,1 mm
    1 * 5 ST
    1 * 5 ST - 2 740,00 €
    2 740,00 €
    1 * 5 ST
       2 740,00 €
    Specifications
    Cat. No.
    PERCN6502012
  • Clarus splitless glass liner
    6,2 mm
    1 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 512,00 €
    512,00 €
    1 * 1 ST
       512,00 €
    Specifications
    Cat. No.
    PERCN6502006
  • Cyclo double gooseneck liner for split operation
    6,2 mm
    4 mm
    92,1
    1 * 1 ST
    1 * 1 ST - 668,00 €
    668,00 €
    1 * 1 ST
       668,00 €
    Specifications
    Cat. No.
    PERCN6502005
  • Glass liner for splitless operation
    6,2 mm
    2 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 93,40 €
    93,40 €
    1 * 1 ST
       93,40 €
    Specifications
    Cat. No.
    PERCN6101372
  • Glass liner for split operation
    6,2 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 81,00 €
    81,00 €
    1 * 1 ST
       81,00 €
    Specifications
    Cat. No.
    PERCN6101052
  • Quartz liner for splitless operation
    6,2 mm
    2 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 53,60 €
    53,60 €
    1 * 1 ST
       53,60 €
    Specifications
    Cat. No.
    PERCN6121002
  • Quartz liner for split operation
    6,2 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 81,00 €
    81,00 €
    1 * 1 ST
       81,00 €
    Specifications
    Cat. No.
    PERCN6121001
  • Siltek deactivated double gooseneck glass liner
    6,2 mm
    2 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 2 690,00 €
    2 690,00 €
    1 * 1 ST
       2 690,00 €
    Specifications
    Cat. No.
    PERCN6502004
  • Siltek deactivated double gooseneck glass liner
    6,2 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 441,00 €
    441,00 €
    1 * 1 ST
       441,00 €
    Specifications
    Cat. No.
    PERCN6502003
  • Split glass liner (with wool)
    6 mm
    4 mm
    92,1 mm
    1 * 5 ST
    1 * 5 ST - 391,00 €
    391,00 €
    1 * 5 ST
       391,00 €
    Specifications
    Cat. No.
    PERCN6502009
  • Split glass liner siltek deactivated surface liner
    6,2 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 660,00 €
    660,00 €
    1 * 1 ST
       660,00 €
    Specifications
    Cat. No.
    PERCN6121020
  • Split siltek deactivated glass liner (with wool)
    6 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 607,00 €
    607,00 €
    1 * 1 ST
       607,00 €
    Specifications
    Cat. No.
    PERCN6502010
  • Splitless liner
    5 mm
    2 mm
    100 mm
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
    Specifications
    Cat. No.
    PERCN6502007
Packed column injector liners
  • Omschrijving
    Ø uitw.
    Ø inw.
    Lengte
  • Cyclo uniliner
    6,2 mm
    4
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 630,00 €
    630,00 €
    1 * 1 ST
       630,00 €
    Specifications
    Cat. No.
    PERCN6502017
  • Drilled uniliner (hole on top)
    6,2 mm
    4 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 4 310,00 €
    4 310,00 €
    1 * 1 ST
       4 310,00 €
    Specifications
    Cat. No.
    PERCN6121022
  • Wide-bore column glass liner
    4 mm
    6 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 93,40 €
    93,40 €
    1 * 1 ST
       93,40 €
    Specifications
    Cat. No.
    PERCN6101375
  • Wide-bore column on/off quartz liner
    4 mm
    6 mm
    92,1 mm
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €
    Specifications
    Cat. No.
    PERCN6121003