Barrel valve
Metalen cilinder met metalen BSP Trisure-schroefdraad 2"
Catalogus # BURK0536-0010
Leverancier: BURKLE
Ga naar:
Product Details & Documents
The barrel valve for opening the barrel closure as well as emptying and reclosing it even with lying, full barrels without having to move them. Also used for highly viscous liquids.
Specificaties
- Voor:Metalen cilinder met metalen BSP Trisure-schroefdraad 2"