Vacuum stopcocks, angled, Witaflo
3 mm
Catalogus # WITG0.290.203
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
High vacuum stopcocks, Witaflo-red are made of borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
With adjustable PTFE-seal, leak rate <10-6, usable between -20 and +200 °C, with overtwist safety device.
Specificaties
- Boring Ø:3 mm