Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), Glass bottle, 1 L
Catalog # MSPPTG10001
Marketsource
Supplier: MARKET SOURCE PART PROCESS
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
Catalog # MSPPTG10001
Supplier: MARKET SOURCE PART PROCESS
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Verpakking:Glass bottle
- VPE:1 L
- Omschrijving:TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)