Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)

Buffer, TRIS-glycine solution 10X concentrate (TG buffer)

TRIS-glycine buffer solution 10X concentrate (TG buffer), Glass bottle, 1 L
Catalogus # BRAD161-0734
Marketsource
Leverancier:   BIO RAD
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)
Catalogus # BRAD161-0734
Leverancier:  BIO RAD
Marketsource
CAS-nummer:  
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)

TRIS-glycine buffer solution 10X concentrate (TG buffer), Glass bottle, 1 L

CAS-nummer:  
Price
61,80 €
Log in voor beschikbaarheid
1 * 1 L
Product Details & Documents
Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    1 L
  • Omschrijving:
    TRIS-glycine buffer solution 10X concentrate (TG buffer)

Specificaties