Orders LinkContactUsLinkComponent
EasyReach™ Cleaning Tools
Supplier:   CONTEC
EasyReach™ Cleaning Tools
Supplier:  CONTEC
Certificates
EasyReach™ Cleaning Tools
EasyReach™ Cleaning Tools
9 Options Available Below
Product Details & Documents
The lightweight, stainless steel EasyReach™ Cleaning Tool is designed for cleaning isolators, laminar air flow hoods, glove boxes, and other mini environments. Rectangular shape makes overlapping straight-line action easy to achieve. Hinged joint and lightweight design makes one-handed cleaning comfortable and improves access to hard-to-reach areas.
  • Rounded tool edges and strong, durable, double-layer pads ensure equipment is not damaged during use
  • Lightweight EasyReach™ Tool can easily be manipulated, is comfortable to use, and minimizes fatigue
  • All components are designed to be chemical resistant, minimizing the risk of contamination from degradation
  • Presaturated option removes the need for 70% IPA spray bottles, reducing inventory and minimizing waste costs
  • Sorbent double layer Quiltec sealed edge polyester cleaning pads ensure particles and fibers are minimized
  • Polyester cleaning pads leave no unwanted surface residues
  • Available validated sterile
The single-use 100% polyester EasyReach™ cleaning pads provide a low level of particulates which is essential for use in product contact areas. The pads are very absorbent, ensuring even coverage of disinfectant or IPA. Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame, but is easy to replace with gloved hands. Sterile pads presaturated with 70% IPA are also available.
Product Family Options
EasyReach™ reinigingsdoekjes
Product Information
  • Omschrijving
    l×b
    Lengte
    Capaciteit
    Verpakt
    Availability
    Price
  • Cleaning pads, EasyReach™, quilted polyester, sewn
    190×71 mm (7.5×2.8")
    4/zak, 15 zakken/doos
    Log in for availability
    Log in for availability
    1 * 60 ST
    1 * 60 ST - 488,00 €
    488,00 €
    1 * 60 ST
       488,00 €
  • Reinigingspads, EasyReach™, 100% gebreid polyester, steriel
    190×71 mm (7.5×2.8")
    4 stuks per zak, 15 zakken per doos
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 60 ST
    1 * 60 ST - 394,00 €
    394,00 €
    Log in for availability
    Each
    Each - 378,00 €
    378,00 €
    1 * 60 ST
       394,00 €
    Each
       378,00 €
  • Cleaning pads, EasyReach™, quilted polyester, ultrasonically sealed
    190×71 mm (7.5×2.8")
    4 each/bag, 25 bags/case
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 730,00 €
    730,00 €
    1 * 100 ST
       730,00 €
  • Cleaning pads, EasyReach™, quilted polyester, sterile, ultrasonically sealed
    190×71 mm (7.5×2.8")
    4 each/bag, 25 bags/case
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 765,00 €
    765,00 €
    1 * 100 ST
       765,00 €
  • Reinigingspads, EasyReach™, 100% gebreid polyester, steriel, voorgedrenkt met 70% IPA en 30% gedemineraliseerd water
    190×71 mm (7.5×2.8")
    8 stuks per zak, 8 zakken per doos
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 64 ST
    1 * 64 ST - 690,00 €
    690,00 €
    Log in for availability
    Each
    Each - 663,00 €
    663,00 €
    1 * 64 ST
       690,00 €
    Each
       663,00 €
EasyReach™ reinigingsgereedschap
Product Information
  • Omschrijving
    l×b
    Lengte
    Capaciteit
    Verpakt
    Availability
    Price
  • QuickConnect™ EasyReach™ lay-flat frame, polypropylene
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,60 €
    19,60 €
    1 * 1 ST
       19,60 €
  • EasyReach™ stainless steel lay flat mop frame
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €
  • EasyReach™ stainless steel one-piece lay flat frame
    420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 210,00 €
    210,00 €
    1 * 1 ST
       210,00 €
  • QuickConnect handle, telescopic, stainless steel
    400 - 760 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 401,00 €
    401,00 €
    1 * 1 ST
       401,00 €