Contact plate and Petri dish reader, SP Bel-Art
Catalog # 710-0079
Supplier: Bel-Art Products, a Part of SP
Specifications
- Omschrijving:Contact plate and petri dish reader
- VPE:1
Specifications
About this item
Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes, this reader allows safe examination of the bottom of plates without risking spills on the face and eyes.
- Ideal for examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
- Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
- Clear deck for resting plates
- Large, adjustable mirror is perfect for viewing plate underside
- Mirror magnification is 3X actual plate size