Orders LinkContactUsLinkComponent
VWR®, Nucleic acid loading dyes
Agarose gel loading dye with 3 tracking dyes and 15% Ficoll (6X), 5 mL
Catalogus # E190-5ML
Leverancier:   VWR Chemicals
VWR®, Nucleic acid loading dyes
Catalogus # E190-5ML
Leverancier:  VWR Chemicals
CAS-nummer:  
VWR®, Nucleic acid loading dyes
VWR®, Nucleic acid loading dyes
Agarose gel loading dye with 3 tracking dyes and 15% Ficoll (6X), 5 mL
CAS-nummer:  
Price
58,90 €
Log in voor beschikbaarheid
1 * 5 mL
Product Details & Documents
Designed to optimise loading nucleic acids in agarose gels, the loading dye imparts colour to the sample to facilitate the loading process and increase the density of the sample to ensure efficient sample distribution into each well. The dye migrates independently from the sample, allowing estimation of the migration of nucleic acids.
  • Eliminates exposure to irritating and harmful powdered dyes
  • Free from DNase, RNase and protease
  • Functionally and analytically tested
Specificaties
Specificaties
  • VPE:
    5 mL
  • Omschrijving:
    Agarose gel loading dye with 3 tracking dyes and 15% Ficoll (6X)