γ-Cyclodextrine
Glass bottle, 5 mL
Catalogus # BECL713350
Marketsource
Leverancier: Beckman Coulter
γ-Cyclodextrine
Catalogus # BECL713350
Leverancier: Beckman Coulter
Marketsource
CAS-nummer: 17465-86-0
Ga naar:
Product Details & Documents
Synoniemen: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- CAS-nummer:17465-86-0
- Formule:C₄₈H₈₀O₄₀
- MDL-nummer:MFCD00009595
- Molecular Weight:1297.1248 g/mol
- EINECS:241-482-4
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:5 mL