Orders LinkContactUsLinkComponent
β-Cyclodextrine
Glass bottle for solids, 100 g
Catalogus # MOLE31870153-100G
Marketsource
Leverancier:   Molekula
β-Cyclodextrine
Catalogus # MOLE31870153-100G
Leverancier:  Molekula
Marketsource
CAS-nummer:  7585-39-9
β-Cyclodextrine
Glass bottle for solids, 100 g
CAS-nummer:  7585-39-9
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
Synoniemen:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS-nummer:7585-39-9
  • Formule:C₄₂H₇₀O₃₅
  • MDL-nummer:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • EINECS:231-493-2
Specificaties
Specificaties
  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g