Order Entry
Belgium
ContactUsLinkComponent
 
Pesticide referentiestandaard, Amitraz 10 µg/ml in cyclohexane
Catalog # EHERL10230000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide referentiestandaard, Amitraz 10 µg/ml in cyclohexane
Catalog # EHERL10230000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    10 mL
  • CAS No.:
    33089-61-1
  • Omschrijving:
    Amitraz 10 µg/ml in cyclohexane
  • Formule:
    C₁₉H₂₃N₃
  • Omschrijving:
    Amitra

Specifications

About this item