Manganese standard solution, 10000 mg/L Mn in 2 M nitric acid (from Mn(NO₃)₂) standard for AAS
Catalog # KRAF14273.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentratie:10000 mg/L
- Matrix:2 M HNO₃
- Verpakking:Plastic bottle
- VPE:100 mL
- Environmentally Preferable:
- Element:Mangaan
- Symbool:Mn