Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) Certipur® standard for AAS, Supelco®
: Merck
Tungsten standard solution, 1000 mg/L W in water (from (NH₄)₂WO₄) Certipur® standard for AAS, Supelco®
: Merck
: Traceable to SRM from NIST
- M-Clarity quality level:MQ100
Frequently Bought Together
Kaliumpermanganaat ≥99%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Nitrilhandschoenen
: VWR Collection
Sodium iodide ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent
: VWR Chemicals
VWR®, gezichtstissues
: VWR Collection
VWR® AVS TITRINORM, Bufferoplossingen, 20 °C
: VWR Chemicals
Manganese standard solution, 10 mg/L Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
: Thermo Fisher Scientific
Nicotinezuur (Niacin)
: PanReac AppliChem