Orders LinkContactUsLinkComponent
Multi-channel gel loading syringes
Leverancier:   HAMILTON BONADUZ
Multi-channel gel loading syringes
Leverancier:  HAMILTON BONADUZ
Certificaten
Multi-channel gel loading syringes
Multi-channel gel loading syringes
8 Opties Hieronder Beschikbaar
Product Details & Documents
Multi-channel gel loading syringes are designed for multiplex DNA sequencing methodologies. They can rapidly transfer 1, 8, or 12 samples to sequencing gels, 96-well plates, or nylon membranes.
  • Dispense from 0,2 to 10 µl of sample per syringe simultaneously
  • 9 mm needle spacing and three needle diameters optimise sample transfers
  • Adjustable volume stop that allows faster, more accurate reloading and excellent reproducibility
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision machined PTFE plunger tip which creates a leak-free seal.
Cemented needle (N): For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable needle (RN): The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length and point style to optimise the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle point style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting.
Product Family Options
Single-channel
Productinformatie
  • Volume
    Lengte
    Ø uitw.
    Tip type
    Type
    Beschikbaarheid
    Price
  • HAMI80081
    Cemented needle multi-channel gel loading syringe, single-channel
    Cemented needle multi-channel gel loading syringe, single-channel
    10 µl
    25,4 mm
    0,2 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • HAMI84505
    Cemented needle multi-channel gel loading syringe, single-channel
    Cemented needle multi-channel gel loading syringe, single-channel
    10 µl
    25,4 mm
    0,3 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • HAMI84504
    Cemented needle multi-channel gel loading syringe, single-channel
    Cemented needle multi-channel gel loading syringe, single-channel
    10 µl
    25,4 mm
    0,4 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 95,20 €
    95,20 €
    1 * 1 ST
       95,20 €
  • HAMI80091
    Removable needle multi-channel gel loading syringe, single-channel
    Removable needle multi-channel gel loading syringe, single-channel
    10 µl
    25,4 mm
    0,2 mm
    3
    1701 RN
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
Multi-channel (8-channel)
Productinformatie
  • Volume
    Lengte
    Ø uitw.
    Tip type
    Type
    Beschikbaarheid
    Price
  • HAMI84511
    Cemented needle multi-channel gel loading syringe, 8-channel
    Cemented needle multi-channel gel loading syringe, 8-channel
    10 µl
    25,4 mm
    0,2 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 1 520,00 €
    1 520,00 €
    1 * 1 ST
       1 520,00 €
  • HAMI84503
    Cemented needle multi-channel gel loading syringe, 8-channel
    Cemented needle multi-channel gel loading syringe, 8-channel
    10 µl
    25,4 mm
    0,3 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • HAMI84502
    Cemented needle multi-channel gel loading syringe, 8-channel
    Cemented needle multi-channel gel loading syringe, 8-channel
    10 µl
    25,4 mm
    0,4 mm
    3
    1701 N
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • HAMI84711
    Removable needle multi-channel gel loading syringe, 8-channel
    Removable needle multi-channel gel loading syringe, 8-channel
    10 µl
    25,4 mm
    0,2 mm
    3
    1701 RN
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 1 660,00 €
    1 660,00 €
    1 * 1 ST
       1 660,00 €
Accessories
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement syringe (barrel incl plunger)
    Replacement syringe (barrel incl plunger)
    HAMI80023
    0,2 mm
    Multi-channel gel loading syringes, for <0,2 mm gel
    Log in voor beschikbaarheid
    1 * 1 ST - 93,10 €
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement syringe (barrel incl plunger)
    Replacement syringe (barrel incl plunger)
    HAMI80021
    0,4 mm
    Multi-channel gel loading syringes, for <0,4 mm gel
    Log in voor beschikbaarheid
    1 * 1 ST - 93,10 €
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement syringe (barrel incl plunger)
    Replacement syringe (barrel incl plunger)
    HAMI80022
    0,3 mm
    Multi-channel gel loading syringes, for <0,3 mm gel
    Log in voor beschikbaarheid
    1 * 1 ST - 95,20 €
Productinformatie
  • Catalogus #
    Voor
    Beschikbaarheid
    Price
  • RN nut
    RN nut
    HAMI30902
    Multi-channel gel loading syringes, for <0,2, 0,3, 0,4 mm gel
    Log in voor beschikbaarheid
    1 * 1 ST - 14,10 €
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement needle
    Replacement needle
    HAMI8651-02
    0,3 mm
    Multi-channel gel loading syringes, for <0,3 mm gel
    Log in voor beschikbaarheid
    1 * 6 ST - 304,00 €
Productinformatie
  • Catalogus #
    Voor
    Beschikbaarheid
    Price
  • Guide rod stop collar
    Guide rod stop collar
    HAMI1993-01
    Multi-channel gel loading syringes
    Log in voor beschikbaarheid
    1 * 2 ST
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Reinigingsdraden, 0,076 mm draad OD, voor 23s gauge naalden
    Reinigingsdraden, 0,076 mm draad OD, voor 23s gauge naalden
    HAMI18306
    Log in voor beschikbaarheid
    1 * 10 ST - 22,80 €
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement needle
    Replacement needle
    HAMI8651-01
    0,2 mm
    Multi-channel gel loading syringes, for <0,2 mm gel
    Log in voor beschikbaarheid
    1 * 6 ST - 304,00 €
Productinformatie
  • Catalogus #
    Ø uitw.
    Voor
    Beschikbaarheid
    Price
  • Replacement needle
    Replacement needle
    HAMI8651-03
    0,4 mm
    Multi-channel gel loading syringes, for <0,4 mm gel
    Log in voor beschikbaarheid
    1 * 6 ST - 309,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.