ContactUsLinkComponent
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Catalogus # A3945.1000
Leverancier:  PanReac AppliChem
CAS-nummer:  
Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
Catalogus # A3945.1000
Leverancier:  PanReac AppliChem
CAS-nummer:  
Specificaties
  • Verpakking:
    Glass bottle
  • VPE:
    1 L
  • Omschrijving:
    Tris-Borate-EDTA (TBE) buffer, 10X, Molecular biology grade
  • Environmentally Preferable:
Specificaties
Over dit artikel
10X geconcenteerde oplossing van 0,89 M tris, 0,89 M boorzuur en 0,02 M EDTA.Na₂, pH 8,3±0,2.
DNases, RNases, and proteases not detectable.