VWR®, folding illuminated magnifiers
Catalog # 630-1091
Supplier: VWR Collection
Specifications
- Ø:17 mm
- Vergroting:10×
- Omschrijving:Folding illuminated magnifier
- Environmentally Preferable:
- VPE:1
Specifications
About this item
These folding aliminium magnifiers with LED illumination are ideal for the professional examination of jewels, fibres, watches and hallmarks.
Colour: Black
Delivery: Supplied in a transparent box with three LR 1130 batteries.