VWR®, folding linen tester
Catalogus # 630-1087
Leverancier: VWR Collection
Specificaties
- Ø:10 mm
- Vergroting:10×
- Omschrijving:Folding magnifier, plastic
- Seq:02
- Environmentally Preferable:
- VPE:1
Specificaties
Over dit artikel
These linen testers are widely used in industrial electronics and laboratories, as well as for photography.
- Glazen lens
- Ledmodel (630-1090)
- Kunststof of aluminium inklapbare modellen
Colour: Black