Orders LinkContactUsLinkComponent
Testplaten voor Eclipse refractometers
Testplaten voor Eclipse refractometers
Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD, 6 mL
Catalogus # 635-0000
Leverancier:   BELLINGHAM STANLEY
Testplaten voor Eclipse refractometers
Catalogus # 635-0000
Leverancier:  BELLINGHAM STANLEY
CAS-nummer:  
Testplaten voor Eclipse refractometers
Testplaten voor Eclipse refractometers
Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD, 6 mL
CAS-nummer:  
Price
61,50 €
Log in voor beschikbaarheid
1 * 6 mL
Product Details & Documents
Solid test plates offer a convenient method of calibration and verifying the performance of optical handheld and Abbe refractometers.
  • Silica test plates for added traceability
  • Glass test plates for honey handheld refractometers
  • No temperature correction required
  • Inert material offering longevity
Depending on the type of instrument and the scale range to be verified, a choice can be made between glass or silica test plates. Silica test plates are traceable to recognised standards and may be recertified as part of a quality control standard operating procedure.
Delivery information: Supplied with a certificate of verification. Additional certification for silica plates is available as an optional extra.
Specificaties
Specificaties
  • VPE:
    6 mL
  • Omschrijving:
    Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD