VWR®, PP-schroefdoppen, ND24 met septum
Leverancier: LA PHA PACK
Certificaten
Over dit artikel
Gerelateerde informatie
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
TLC- en GLP-platen, kiezelgel 60, ongemodifiëerd, glazen platen
Leverancier: Merck
VWR®, Screw Caps for Short Thread Vials ND9
Leverancier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
VWR®, Short Thread Vials ND9
Leverancier: VWR Collection
VWR®, Snap Cap Vials
Leverancier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Cork rings for round bottom flasks
Leverancier: GLASWARENFABRIK KARL HECHT
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Leverancier: VWR Chemicals
VWR®, Bekers, lage vorm
Leverancier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals