Bellow connectors
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
- Universal chemical resistance
- Suitable for vacuum applications
- Temperature resistance from −200 to +250 °C
- Compensates vibrations from vacuum pumps
- Suiltable for length compensation of heated columns and angular misalignment
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
- Universal chemical resistance
- Suitable for vacuum applications
- Temperature resistance from −200 to +250 °C
- Compensates vibrations from vacuum pumps
- Suiltable for length compensation of heated columns and angular misalignment
Product Family Options
Sharp folds, 120° deflection
- OmschrijvingAfmetingen kernTotale lengteHuls
Round folds, 40° deflection
- OmschrijvingAfmetingen kernTotale lengteHuls