Accessories for SCAT Politainer
Catalog # SCTV107338
Supplier: S.C.A.T.
Specifications
- Voor:Foldable politainer with GL 38
- Omschrijving:Thread adapter, PTFE, GL 38 on GL 45
- Seq:04
- VPE:1
Specifications
About this item
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
- Thread to adjust the thread from GL38 to GL45
- Especially suitable for SCAT Politainer
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.