Burette and thermometer clamps with bosshead, AUTOMATIC
Catalog # BOCH5625
Supplier: BOCHEM
Specifications
- Voor:Buretten en thermometers
- Spanwijdte:0 - 25 mm
- VPE:1
Specifications
About this item
These chromium plated clamps are designed for holding burettes or thermometers (Ø 0 - 25 mm). The bosshead (Ø 13,5 mm) can be rotated and locked at an angle of 90°.