Celite® Standard Super-cel, filter aid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91053-39-3
:61790-53-2
MDL Number:MFCD00132803
Merck Index:14,4973
Boiling Pt:2200 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.3 g/cm³ (20 °C)
EINECS:319-127-6
:GHS08
:GHS07
Storage Temperature:Kamertemperatuur
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together