2,3-Dibromothiophene
Glass bottle for solids, 5 g
Catalogus # MOLE34038529-5G
Marketsource
Leverancier: Molekula
2,3-Dibromothiophene
Catalogus # MOLE34038529-5G
Leverancier: Molekula
Marketsource
CAS-nummer: 3140-93-0
Ga naar:
Product Details & Documents
- CAS-nummer:3140-93-0
- Formule:C₄H₂Br₂S
- MDL-nummer:MFCD00005418
- Molecular Weight:241.93 g/mol
- Kokend Pt:130 °C (1013 hPa)
- Smelten Pt:–17,5 °C
- Dichtheid:2,2 g/cm³ (20 °C)
- EINECS:221-542-6
- UN:1993
- ADR:3,III
- :Density: 2.2 g/cm³ (20 °C)
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:5 g