Orders LinkContactUsLinkComponent
Adapters, receiver, BOLA
19/26 NS, 29/32 NS
Catalogus # BOHLB292-02
Leverancier:   Bohlender
Adapters, receiver, BOLA
Catalogus # BOHLB292-02
Leverancier:  Bohlender
Adapters, receiver, BOLA
Adapters, receiver, BOLA
19/26 NS, 29/32 NS
Price
254,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
Ground joint cone and socket size NS 29, as well as socket size NS 19 for example for vacuum stopcock (see Cat. No. B 293-02). Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    19/26 NS