Sampling pumps, BOLA
Monsternamepomp, met GL 45-schroefdraad, voor flessen met 100 - 2000 liter, PP/PTFE, 50 ml/stroke, 6 mm
Catalogus # BOHLA124-16
Leverancier: Bohlender
Ga naar:
Product Details & Documents
Sampling pump is made up of PP and PTFE.
- Compact construction and easy handling
- Quick and low-risk pumping of ultrapure liquids
- Ergonomic and usable anywhere
- Universal chemical resistance
A pull on the ball handle produces a slight vacuum in the sampling bottle. Due to this vacuum, the sample is sucked into the sampling bottle.
Glass bottles and plastic bottles with a GL 45 thread can be used as sampling bottles.
Applicable for pumping liquids from sources that are not easily accessible; also, suitable for liquids with a high viscosity like oils.
Delivery information: Supplied with tubing outer Ø (6 mm), PTFE tubing (2 m).
Specificaties
- Stroomsnelheid:50 ml/stroke
- Omschrijving:Monsternamepomp, met GL 45-schroefdraad, voor flessen met 100 - 2000 liter
- Materiaal:PP/PTFE
- Buis Ø:6 mm