Potassium cyanide
Glass bottle, 500 mL
Leverancier: PROLABO CMP
Ga naar:
Product Details & Documents
- CAS-nummer:151-50-8
- Formule:KCN
- MDL-nummer:MFCD00011397
- Molecular Weight:65.12 g/mol
- Merck index:13,07709
- Kokend Pt:1625 °C (1013 hPa)
- Smelten Pt:634 °C
- Dichtheid:1,55 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:205-792-3
- UN:1680
- ADR:6.1,I
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- Verpakking:Glass bottle
- VPE:500 mL
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



