1,4-Dioxaan, LiChrosolv® for HPLC, Supelco®
: Merck
:123-91-1
:
:C₄H₈O₂
:MFCD00006571
:88.11 g/mol
:13,03330
:101.1 °C (1013 hPa)
:11.8 °C
:1.03 g/cm³ (20 °C)
:204-661-8
:1165
:3,II
:M-Clarity™ quality level = MQ100
:12 °C
:GHS02
:GHS08
:GHS07
:Kamertemperatuur
Frequently Bought Together