Natriumhydroxide concentrate for dilution to 2L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:120 °C
Density:1,350 g/cm³
Storage Temperature:Kamertemperatuur
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together