Flexible couplings for stirrers
Catalogus # LENZ06400000
Leverancier: Lenz Laborglas GmbH & CO.KG
Specificaties
- Omschrijving:Flexible coupling for stirrers, with pivot 8 - 10 mm
- VPE:1
Specificaties
Over dit artikel
Flexible coupling made of stainless steel.
- Compensates for axial and height differences
- No adjustments necessary for motor and stirring rod
- Stainless steel clamping chuck with clamping range 8 to 10 mm