Order Entry
Belgium
ContactUsLinkComponent
o-Xyleen ≥98.0%, Reagent Grade
o-Xyleen ≥98.0%, Reagent Grade
Catalogus # X1040-18L
Leverancier:  Honeywell Chemicals
CAS-nummer:  
undefined
o-Xyleen ≥98.0%, Reagent Grade
Catalogus # X1040-18L
Leverancier:  Honeywell Chemicals
CAS-nummer:  

Specificaties

  • Verpakking:
    Stainless steel can
  • VPE:
    18 L
  • Seq:
    5267

Specificaties

Specificatie testresultaten

  • Vapor Density:
    3.7 (vs air)
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 98.0 %
  • m-, p-Xylene (GC):
    Max. 1.0 %
  • Ethyl benzene (GC):
    Max. 0.50 %
  • Toluene (GC):
    Max. 0.50 %
  • Autoignition Temperature:
    867 °F
  • Explosion Limit:
    7 %
  • Refractive Index:
    n20/D 1.505 (lit.)

Specificatie testresultaten

Over dit artikel

 :
 :106.17 g/mol
Merck index:12,10214
Kokend Pt:7.6 %(V)
Smelten Pt:-25 °C
Smelten Pt:–25 °C
Dichtheid:0.88 g/cm³ (20 °C)
Flits Pt:30 °C
EINECS:202-422-2
UN:1307
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
Temperatuur bij opslag:Kamertemperatuur
CAS-nummer:95-47-6
Formule:C₈H₁₀
MDL-nummer:MFCD00008519