Thermometer clamp, with swivelling bosshead
Catalogus # BOCH5621
Leverancier: BOCHEM
Specificaties
- Voor:Thermometers
- Spanwijdte:6 - 15 mm
- VPE:1
Specificaties
Over dit artikel
These chromium plated clamps are used for holding thermometers (Ø 6 to 15 mm). Swivelling bosshead (Ø 16,5 mm) can be rotated and locked at any angle (0 to 360°).