Orders LinkContactUsLinkComponent
Beaker tongs, stainless steel
Beaker tongs, stainless steel
Leverancier:   BOCHEM
Beaker tongs, stainless steel
Leverancier:  BOCHEM
Certificaten
Beaker tongs, stainless steel
Beaker tongs, stainless steel
2 Opties Hieronder Beschikbaar
Product Details & Documents
These beaker tongs are made of electrolytically polished stainless steel. The jaws are covered with silicone tubing.
Product Family Options
Productinformatie
  • Lengte
    Spanbereik
    Beschikbaarheid
    Price
  • 250 mm
    25 - 100 mm
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 23,00 €
    23,00 €
    1 * 1 ST
       23,00 €
  • 250 mm
    55 - 100 mm
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 20,20 €
    20,20 €
    1 * 1 ST
       20,20 €