Klemmen, met vier pinnen, universeel
Catalog # RSGA400.000
Supplier: RSG
Specifications
- Spanwijdte:90 mm
- VPE:1
Specifications
About this item
Universal clamp made from zinc die-cast.
- Four-finger jaws
- Jaws fitted with cork
Universal clamp made from zinc die-cast.