General purpose spoons
: RSG
Double-ended spoons made from stainless steel.
- Round handle (Ø 5 mm)
- One spatula end, one spoon end
Frequently Bought Together
Maatbekers met handgreep, PP
: BURKLE
Pincetten voor dekglaasjes
: USBECK Laborgeräte
VWR® Standaardlijntransferpipetten
: VWR Collection
Zwavelzuur 40%, Supelco®
: Merck
Screw Caps for Bottles
: RIXIUS
VWR®, Wegwerpspatels, PP
: VWR Collection
Objectglaasjes
: MARIENFELD
Trechters met korte steel, DURAN®
: DWK Life Sciences
Laboratoriumlepel
: RSG
DuPont™ AmberLite™ IRN77 H cation exchange resin
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Zuurbestendige handschoenen, Camapren®
: KCL by Honeywell
Magnetic stirrers, boxed sets
: COWIE
Zoutzuur 32%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Wissers
: DELTALAB
Squalene ≥98%
: Thermo Fisher Scientific
General purpose spoons
: RSG