2,3,4-Trichloorfenol
Catalog # EHERC17773400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Verpakking:Glass bottle for solids
- VPE:25 mg
- Seq:2315
Specifications
About this item
:
:197.45 g/mol
Boiling Pt:256.8 °C (1013 hPa)
Melting Pt:77…79 °C
EINECS:240-083-2
UN:2020
ADR:6.1,III
CAS Number:15950-66-0
Formula:C₆H₃Cl₃O