Spuitflessen
1000 ml, 271 mm, 90 mm
Catalogus # 215-4419
Leverancier: KARTELL
Ga naar:
Product Details & Documents
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
- Suitable for foodstuff
Spray bottle atomiser with adjustment jet.
Documents
Specificaties
- Ø uitw.:90 mm
- Capaciteit:1000 ml
- Hoogte:271 mm