Sample concentrators, Vivaspin®
About this item
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
- One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
- Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
- Vertical PES membrane minimises membrane blockage and tolerates high flow rates
- Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
24 Options Available Below
Product Details & Documents
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
- One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
- Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
- Vertical PES membrane minimises membrane blockage and tolerates high flow rates
- Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Documents
Certificates
Search by Lot Number28932218_DATA FILE 28932225_DATA FILE 28932235_DATA FILE 28932236_DATA FILE 28932237_DATA FILE
Datasheet
28932218_HAND BOOK 28932218_INSTRUCTIONS 28932225_HAND BOOK 28932225_INSTRUCTIONS 28932235_HAND BOOK
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications for Product Family
Specifications
- Catalog No:
- 28-9322-18
- 28-9322-23
- 28-9322-25
- 28-9322-35
- 28-9322-36
- 28-9322-37
- 28-9322-40
- 28-9322-45
- 28-9322-47
- 28-9322-48
- 28-9322-57
- 28-9322-58
- 28-9322-93
- 28-9322-94
- 28-9322-96
- 28-9323-17
- 28-9323-18
- 28-9323-19
- 28-9323-58
- 28-9323-59
- 28-9323-60
- 28-9323-61
- 28-9323-62
- 28-9323-63
- Type:
- Vivaspin® 500
- Vivaspin® 500
- Vivaspin® 500
- Vivaspin® 500
- Vivaspin® 500
- Vivaspin® 500
- Vivaspin® 2
- Vivaspin® 2
- Vivaspin® 2
- Vivaspin® 2
- Vivaspin® 2
- Vivaspin® 2
- Vivaspin® 6
- Vivaspin® 6
- Vivaspin® 6
- Vivaspin® 6
- Vivaspin® 6
- Vivaspin® 6
- Vivaspin® 20
- Vivaspin® 20
- Vivaspin® 20
- Vivaspin® 20
- Vivaspin® 20
- Vivaspin® 20
- Filtratieoppervlak:
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 0,5 cm²
- 1,2 cm²
- 1,2 cm²
- 1,2 cm²
- 1,2 cm²
- 1,2 cm²
- 1,2 cm²
- 2,5 cm²
- 2,5 cm²
- 2,5 cm²
- 2,5 cm²
- 2,5 cm²
- 2,5 cm²
- 6,0 cm²
- 6,0 cm²
- 6,0 cm²
- 6,0 cm²
- 6,0 cm²
- 6,0 cm²
- Dood volume:
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- <10 µl
- 50 µl
- 50 µl
- 50 µl
- 50 µl
- 50 µl
- 50 µl
- Ø×H:
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 11×50 mm
- 17×126 mm
- 17×126 mm
- 17×126 mm
- 17×126 mm
- 17×126 mm
- 17×126 mm
- 16×122 mm
- 16×122 mm
- 16×122 mm
- 16×122 mm
- 16×122 mm
- 16×122 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
Specifications
Product Family Options
Vivaspin® 500
Product Information
- OmschrijvingMWCOAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.28-9322-18TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Cat. No.28-9322-23TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Cat. No.28-9322-25TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Cat. No.28-9322-35TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Cat. No.28-9322-36TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm - Specifications:
More
Specifications
Cat. No.28-9322-37TypeVivaspin® 500Filtratieoppervlak0,5 cm²Dood volume<5 µlØ×H11×50 mm
Vivaspin® 2
Product Information
- OmschrijvingMWCOAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.28-9322-40TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm - Specifications:
More
Specifications
Cat. No.28-9322-45TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm - Specifications:
More
Specifications
Cat. No.28-9322-47TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm - Specifications:
More
Specifications
Cat. No.28-9322-48TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm - Specifications:
More
Specifications
Cat. No.28-9322-57TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm - Specifications:
More
Specifications
Cat. No.28-9322-58TypeVivaspin® 2Filtratieoppervlak1,2 cm²Dood volume<10 µlØ×H17×126 mm
Vivaspin® 6
Product Information
- OmschrijvingMWCOAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.28-9322-93TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm - Specifications:
More
Specifications
Cat. No.28-9322-94TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm - Specifications:
More
Specifications
Cat. No.28-9322-96TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm - Specifications:
More
Specifications
Cat. No.28-9323-17TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm - Specifications:
More
Specifications
Cat. No.28-9323-18TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm - Specifications:
More
Specifications
Cat. No.28-9323-19TypeVivaspin® 6Filtratieoppervlak2,5 cm²Dood volume<10 µlØ×H16×122 mm
Vivaspin® 20
Product Information
- OmschrijvingMWCOAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.28-9323-58TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm - Specifications:
More
Specifications
Cat. No.28-9323-59TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm - Specifications:
More
Specifications
Cat. No.28-9323-60TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm - Specifications:
More
Specifications
Cat. No.28-9323-61TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm - Specifications:
More
Specifications
Cat. No.28-9323-62TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm - Specifications:
More
Specifications
Cat. No.28-9323-63TypeVivaspin® 20Filtratieoppervlak6,0 cm²Dood volume50 µlØ×H30×116 mm