Table clamp
Aluminiumlegering, 26 mm, Staven en vierkante staven
Catalogus # USBE2051
Leverancier: USBECK Laborgeräte
Ga naar:
Product Details & Documents
Large table clamp made of powder-coated aluminium alloy.
- For rods and square rods up to 26 mm Ø
- For benches up to 55 mm
Specificaties
- Voor:Staven en vierkante staven
- Spanwijdte:26 mm
- Materiaal:Aluminiumlegering