Organische referentiestandaard, 2-Nitronaftaleen 10 µg/ml in cyclohexaan
EHERL20965200CY
:
- Verpakking:Glass bottle
- VPE:10 mL
- CAS No.:581-89-5
- Omschrijving:2-Nitronaftaleen 10 µg/ml in cyclohexaan
- Formule:C₁₀H₇NO₂
- Omschrijving:2-Nitronaphthalene