Tapes, plastilab®
Tape, 12,0 mm
Catalogus # KART1053
Leverancier: KARTELL
Ga naar:
Product Details & Documents
This tape is made of PTFE.
Length: 12,0 m
Colour: White
Specificaties
- Breedte:12,0 mm
- Omschrijving:Tape