Afsluitkranen, driewegversie, Czako
Catalog # LENZ02631804
Supplier: Lenz Laborglas GmbH & CO.KG
Specifications
- Boring Ø:2,5 mm
- Omschrijving:Stopcock, three-way, with PTFE plug
- VPE:1
- Kraan:18,8 NS
Specifications
About this item
Stopcocks, made of DURAN® borosilicate glass.
- Good temperature resistance
- Good chemical resistance
With PTFE plug and screwthread retaining device.
Certifications: Leakage rate according to DIN 12540-1.