Septumdoppen, Nalgene®
38 - 430, PP
Leverancier: Thermo Fisher Scientific
Septumdoppen, Nalgene®
Catalogus # 391-0073
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Polypropylene closure with thermoplastic elastomer septum.
- Use with 18-gauge or smaller needles
For Nalgene bottles with 38-430 threaded mouth.
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Sluiting diam:38 - 430
- Materiaal:PP
- Cat. No.:391-0073
- Milieu certificering:NAR Certificate of Retirement
- Environmentally Preferable:Lage impact op de fabricage