Membraanschijffilters
Over dit artikel
Gelatine membrane filters in conjunction with the MD8 air samplers (gelatine filter method) are used for the collection of airborne microbes and viruses. These filters are completely water-soluble, therefore, microbes in one sample can be cultivated in different nutrient media or low and high bacteria counts can be measured.
- Pre-sterilised and ready to use
- Maintains viability of the collected microorganisms
- Solubility of the filter is a prerequisite for virus sampling
'Absolute' retention rate of 99,9995% for Bacillus subtilis and niger spores and 99,94% for T3 phages.
6 Opties Hieronder Beschikbaar
Product Details & Documents
Gelatine membrane filters in conjunction with the MD8 air samplers (gelatine filter method) are used for the collection of airborne microbes and viruses. These filters are completely water-soluble, therefore, microbes in one sample can be cultivated in different nutrient media or low and high bacteria counts can be measured.
- Pre-sterilised and ready to use
- Maintains viability of the collected microorganisms
- Solubility of the filter is a prerequisite for virus sampling
'Absolute' retention rate of 99,9995% for Bacillus subtilis and niger spores and 99,94% for T3 phages.
Specifications for Product Family
Specificaties
- Catalog No:
- SART12602-37-ALK
- SART12602-47-ALK
- SART12602-47-ALN
- SART12602-50-ALK
- SART12602-50-ALN
- 512-8500
- Poriëngrootte:
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- 3,00 µm
- Dikte:
- 250 µm
- 250 µm
- 250 µm
- 250 µm
- 250 µm
- 250 µm
- Luchtdebiet:
- 2,7 ml/min/cm² (0,05 bar)
- 2,7 ml/min/cm² (0,05 bar)
- 2,7 ml/min/cm² (0,05 bar)
- 2,7 ml/min/cm² (0,05 bar)
- 2,7 ml/min/cm² (0,05 bar)
- 2,7 l/min/cm² (0,05 bar)
- Maximumtemperatuur:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- Ø:
- 37 mm
- 47 mm
- 47 mm
- 50 mm
- 50 mm
- 80 mm
Specificaties
Product Family Options
Productinformatie
- ØBeschikbaarheidPrice
- Specificaties:
More
Specifications
Cat. No.SART12602-37-ALKPoriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 ml/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ37 mm - Specificaties:
More
Specifications
Cat. No.SART12602-47-ALKPoriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 ml/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ47 mm - Specificaties:
More
Specifications
Cat. No.SART12602-47-ALNPoriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 ml/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ47 mm - Specificaties:
More
Specifications
Cat. No.SART12602-50-ALKPoriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 ml/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ50 mm - Specificaties:
More
Specifications
Cat. No.SART12602-50-ALNPoriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 ml/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ50 mm - Specificaties:
More
Specifications
Cat. No.512-8500Poriëngrootte3,00 µmDikte250 µmLuchtdebiet2,7 l/min/cm² (0,05 bar)Maximumtemperatuur60 °CØ80 mm