Orders LinkContactUsLinkComponent
Potassium acetate, Millipore®
Potassium acetate, Millipore®
Potassium acetate, Millipore®
Certificates
Potassium acetate, Millipore®
Potassium acetate, Millipore®
2 Options Available Below
Product Details & Documents
Synonyms:  Acetic acid potassium salt
  • CAS Number:127-08-2
  • Formula:CH₃COOK
  • MDL Number:MFCD00012458
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,07687
  • Melting Pt:304 °C
  • Density:1,57 g/cm³ (25 °C)
  • Flash Pt:>250 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:204-822-2
Documents
Product Family Options
Product Information
  • Verpakking
    VPE
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 90,50 €
    90,50 €
    1 * 250 g
       90,50 €
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 206,00 €
    206,00 €
    1 * 1 kg
       206,00 €