Order Entry
Belgium
ContactUsLinkComponent
Pesticide referentiestandaard, 2,5-Dichloorfenol 10 µg/ml in methanol
Pesticide referentiestandaard, 2,5-Dichloorfenol 10 µg/ml in methanol
Catalog # EHERL12452000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide referentiestandaard, 2,5-Dichloorfenol 10 µg/ml in methanol
Catalog # EHERL12452000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    10 mL
  • CAS No.:
    583-78-8
  • Omschrijving:
    2,5-Dichloorfenol 10 µg/ml in methanol
  • Formule:
    C₆H₄Cl₂O
  • Omschrijving:
    2,5-Dichlorophenol

Specifications

About this item