Pesticide referentiestandaard, 3,5-Dichloorfenol 10 µg/ml in methanol
Catalog # EHERL12453500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Verpakking:Glass bottle
- VPE:10 mL
- CAS No.:591-35-5
- Omschrijving:3,5-Dichloorfenol 10 µg/ml in methanol
- Formule:C₆H₄Cl₂O
- Omschrijving:3,5-Dichlorophenol