Kaliumchromaat 10% in aqueous solution analytical reagent
:7789-00-6
:
:K₂CrO₄
:MFCD00011368
:1000
:194.19 g/mol
:985
:6.1 III
:12,07784
:232-140-5
:3287
:GHS08
:GHS07
:GHS09
- Environmentally Preferable:
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals