Antibacterial lotion and foam soaps, Stellisept® med
Catalogus # BODL9810200
Leverancier: Bode
Specificaties
- Omschrijving:Antiseptic foam soap, Stellisept® med foam, squeeze bottle
- Seq:03
- VPE:10 L
Specificaties
Over dit artikel
Stellisept® med is an antiseptic wash lotion that can be used for whole-body washing, hygienic handwashing and pre operative body washing.
- Skin-friendly pH value of approximately 5,5
- Contains no alcohol or colourants
- Skin nurturing properties
- Bactericidal activity
Let op: * These articles are only available in Germany.