Lugol's iodine solution
Supplier: Roth Carl
Certificates
About this item
:
Density:1,01 g/cm³ (20 °C)
Storage Temperature:Kamertemperatuur
Frequently Bought Together
Natriumhydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH-indicatiestrips, MQuant®, Supelco®
Supplier: Merck
Testsets COD-cuvettentest voor Spectroquant®, Supelco®
Supplier: Merck
Testsets, Stikstoftest voor Spectroquant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Testsets, ammoniumtest, voor Spectroquant®, Supelco®
Supplier: Merck
pH-teststrips, MQuant®, Supelco®
Supplier: Merck
Teststrips, perzuurtest, MQuant™, Supelco®
Supplier: Merck
Teststrips, chloortest (vrije chloor), MQuant™, Supelco®
Supplier: Merck
Natriumhydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Testsets, fosfaattest, voor Spectroquant®, Supelco®
Supplier: Merck
Teststrips, peroxidetest, MQuant™, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach